Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Man2b2
Entrez Gene ID
17160
Gene Name
mannosidase 2, alpha B2
Gene Aliases
mKIAA0935
Chromosome Location
Chr.5: 36806813 - 36830649 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008550.2 NP_032576.2 5 - 6 755 56
GenBank mRNA AB006458.1 - 5 - 6 733 56
AK004900.1 - 5 - 6 709 56
AK040388.1 - 5 - 6 716 56
AK079969.1 - 5 - 6 737 56
AK151873.1 - 5 - 6 735 56
AK152349.1 - 5 - 6 735 56
AK165376.1 - 5 - 6 748 56
BC066211.1 - 5 - 6 706 56

View Full Product Details