The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139064.2 | NP_620703.1 | 1 - 2 | 360 | 77 |
GenBank mRNA | AJ304865.1 | - | 1 - 2 | 360 | 77 |
AK160165.1 | - | 1 - 2 | 318 | 77 | |
BC052083.1 | - | 1 - 2 | 343 | 77 |