Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbwd1
Entrez Gene ID
226043
Gene Name
COBW domain containing 1
Gene Aliases
AV349248, BC018472
Chromosome Location
Chr.19: 24919916 - 24961616 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146097.3 NP_666209.1 12 - 13 956 90
NR_033744.1 12 - 13 1023 90
GenBank mRNA AK132291.1 - 12 - 13 942 90
BC018472.1 - 12 - 13 898 90
BC027643.1 - 1 - 2 47 90
BC053745.1 - 12 - 13 991 90

View Full Product Details