Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cbwd1
Entrez Gene ID
226043
Gene Name
COBW domain containing 1
Gene Aliases
AV349248, BC018472
Chromosome Location
Chr.19: 24919916 - 24961616 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146097.3 NP_666209.1 5 - 6 562 120
NR_033744.1 5 - 6 629 120
GenBank mRNA AK076027.1 - 5 - 6 517 120
AK132291.1 - 5 - 6 548 120
BC018472.1 - 5 - 6 504 120
BC053745.1 - 5 - 6 597 120

View Full Product Details