The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048192.2 | NP_001041657.1 | 12 - 13 | 2300 | 75 |
NM_001294262.1 | NP_001281191.1 | 11 - 12 | 2328 | 75 | |
NR_122116.1 | 11 - 12 | 2284 | 75 | ||
NR_122117.1 | 11 - 12 | 2284 | 75 | ||
GenBank mRNA | AK034894.1 | - | 11 - 12 | 2306 | 75 |
AK035680.1 | - | 9 - 10 | 1785 | 75 | |
AK083656.1 | - | 10 - 11 | 2472 | 75 | |
BC057349.1 | - | 11 - 12 | 2218 | 75 | |
DQ867034.1 | - | 11 - 12 | 2213 | 75 | |
DQ867035.1 | - | 11 - 12 | 2213 | 75 | |
DQ867036.1 | - | 12 - 13 | 2300 | 75 | |
DQ867037.1 | - | 11 - 12 | 2213 | 75 |