The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080710.2 | NP_001074179.1 | 2 - 3 | 429 | 67 |
GenBank mRNA | AK076465.1 | - | 2 - 3 | 410 | 67 |
BC125450.1 | - | 2 - 3 | 429 | 67 | |
BC125452.1 | - | 2 - 3 | 429 | 67 |