Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chdh
Entrez Gene ID
218865
Gene Name
choline dehydrogenase
Gene Aliases
AU041472, D630034H06Rik
Chromosome Location
Chr.14: 30009000 - 30040466 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136240.1 NP_001129712.1 1 - 2 240 51
GenBank mRNA AK052715.1 - 1 - 2 218 51
AK164042.1 - 1 - 2 76 51
BY773937.1 - 1 - 2 241 51

View Full Product Details