Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abcb5
Entrez Gene ID
77706
Gene Name
ATP-binding cassette, sub-family B (MDR/TAP), member 5
Gene Aliases
9230106F14Rik
Chromosome Location
Chr.12: 118867486 - 118966421 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029961.2 NP_084237.1 8 - 9 891 82
GenBank mRNA AY766239.1 - 8 - 9 855 82

View Full Product Details