The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081291.1 | NP_001074760.1 | 18 - 19 | 3186 | 85 |
GenBank mRNA | BC076600.1 | - | 9 - 10 | 2070 | 85 |
BC151001.1 | - | 18 - 19 | 3159 | 85 | |
BC151009.1 | - | 18 - 19 | 3159 | 85 | |
BC157946.1 | - | 18 - 19 | 3159 | 85 |