The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195084.1 | NP_001182013.1 | 8 - 9 | 1313 | 91 |
NM_008880.3 | NP_032906.2 | 7 - 8 | 1308 | 91 | |
GenBank mRNA | AF015790.1 | - | 8 - 9 | 1079 | 91 |
AK002410.1 | - | 7 - 8 | 1195 | 91 | |
BC002012.1 | - | 8 - 9 | 1179 | 91 | |
BC011299.1 | - | 7 - 8 | 1190 | 91 |