The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204371.1 | NP_001191300.1 | 1 - 2 | 177 | 62 |
GenBank mRNA | BC144731.1 | - | 1 - 2 | 113 | 62 |
S81111.1 | - | 1 - 2 | 58 | 62 |