The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037997.3 | NP_001033086.2 | 11 - 12 | 1663 | 64 |
NM_001286415.1 | NP_001273344.1 | 10 - 11 | 1491 | 64 | |
NM_008000.2 | NP_032026.2 | 6 - 7 | 945 | 64 | |
GenBank mRNA | AK082799.1 | - | 8 - 9 | 1156 | 64 |
AK157828.1 | - | 11 - 12 | 1627 | 64 | |
BC051249.1 | - | 3 - 4 | 407 | 64 | |
BC058100.1 | - | 10 - 11 | 1491 | 64 | |
M32054.1 | - | 6 - 7 | 945 | 64 | |
U76762.1 | - | 10 - 11 | 1596 | 64 |