Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cry1
Entrez Gene ID
12952
Gene Name
cryptochrome 1 (photolyase-like)
Gene Aliases
AU020726, AU021000, Phll1
Chromosome Location
Chr.10: 85131700 - 85185054 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007771.3 NP_031797.1 6 - 7 1398 101
GenBank mRNA AB000777.1 - 6 - 7 834 101
AF156986.1 - 6 - 7 1133 101
AK133818.1 - 6 - 7 1410 101
AK162460.1 - 6 - 7 1398 101
BC022174.1 - 6 - 7 1169 101
BC085499.1 - 6 - 7 1398 101

View Full Product Details