The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011163.4 | NP_035293.1 | 10 - 11 | 1021 | 91 |
GenBank mRNA | AK028602.1 | - | 10 - 11 | 1021 | 91 |
BC016422.1 | - | 10 - 11 | 984 | 91 | |
M65029.1 | - | 10 - 11 | 973 | 91 | |
M93567.1 | - | 10 - 11 | 91 |