The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010553.4 | NP_034683.1 | 12 - 13 | 1809 | 74 |
GenBank mRNA | AF077347.1 | - | 12 - 13 | 1818 | 74 |
BC120598.1 | - | 12 - 13 | 1816 | 74 | |
BC120600.1 | - | 12 - 13 | 1816 | 74 |