The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008451.2 | NP_032477.2 | 15 - 16 | 2081 | 82 |
NR_045528.1 | 15 - 16 | 2113 | 82 | ||
GenBank mRNA | AK141439.1 | - | 15 - 16 | 2006 | 82 |
BC014845.1 | - | 15 - 16 | 1925 | 82 | |
CX565742.1 | - | 7 - 8 | 681 | 82 |