Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bpifb6
Entrez Gene ID
228796
Gene Name
BPI fold containing family B, member 6
Gene Aliases
Bpil3, Gm119, Lplunc6
Chromosome Location
Chr.2: 153900388 - 153912795 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199303.2 NP_955007.2 15 - 16 1379 66
GenBank mRNA AK087711.1 - 4 - 5 233 66
BC048083.1 - 15 - 16 1379 66

View Full Product Details