The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252448.1 | NP_001239377.1 | 2 - 3 | 218 | 98 |
GenBank mRNA | AK138358.1 | - | 2 - 3 | 219 | 98 |
BC052063.1 | - | - | 238 | 98 |