The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286006.1 | NP_001272935.1 | 15 - 16 | 2303 | 84 |
NM_172569.4 | NP_766157.1 | 15 - 16 | 2342 | 84 | |
GenBank mRNA | AK046395.1 | - | 15 - 16 | 2337 | 84 |
AK077635.1 | - | 15 - 16 | 2344 | 84 | |
AK078095.1 | - | 15 - 16 | 2294 | 84 | |
AK129438.1 | - | 15 - 16 | 2320 | 84 | |
AK134361.1 | - | 15 - 16 | 2423 | 84 | |
BC003195.1 | - | 12 - 13 | 1788 | 84 | |
BC005545.1 | - | 2 - 3 | 349 | 84 | |
BC033048.1 | - | 3 - 4 | 542 | 84 | |
BC039964.1 | - | 15 - 16 | 2304 | 84 | |
BC054452.1 | - | 15 - 16 | 2279 | 84 |