The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109905.2 | NP_001103375.1 | 9 - 10 | 1572 | 80 |
NM_001109906.2 | NP_001103376.1 | 9 - 10 | 1590 | 80 | |
NM_001291160.1 | NP_001278089.1 | 9 - 10 | 1572 | 80 | |
NM_011490.4 | NP_035620.1 | 9 - 10 | 1566 | 80 | |
NR_111895.1 | 9 - 10 | 1239 | 80 | ||
GenBank mRNA | AF061942.1 | - | 9 - 10 | 1566 | 80 |
AF395842.1 | - | 8 - 9 | 1293 | 80 | |
AK005002.2 | - | 9 - 10 | 1513 | 80 | |
AK139532.1 | - | 9 - 10 | 1519 | 80 | |
AK145826.1 | - | 9 - 10 | 1557 | 80 | |
AK151835.1 | - | 9 - 10 | 1378 | 80 | |
AK152269.1 | - | 9 - 10 | 1384 | 80 | |
AK152744.1 | - | 9 - 10 | 1378 | 80 | |
AK158952.1 | - | 4 - 5 | 2153 | 80 | |
AK159602.1 | - | 9 - 10 | 1507 | 80 | |
AK160979.1 | - | 9 - 10 | 1531 | 80 | |
AK167637.1 | - | 9 - 10 | 1513 | 80 | |
AY391773.1 | - | 9 - 10 | 1238 | 80 | |
BC012959.1 | - | 9 - 10 | 1536 | 80 | |
BC082277.1 | - | 9 - 10 | 1509 | 80 |