The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053200.2 | NP_444430.2 | 1 - 2 | 140 | 88 |
GenBank mRNA | AB023631.1 | - | 1 - 2 | 100 | 88 |
AF378751.1 | - | 1 - 2 | 115 | 88 | |
AK078879.1 | - | 1 - 2 | 118 | 88 | |
BC019198.1 | - | 1 - 2 | 112 | 88 |