Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif9
Entrez Gene ID
16578
Gene Name
kinesin family member 9
Gene Aliases
-
Chromosome Location
Chr.9: 110476786 - 110525174 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163569.1 NP_001157041.1 7 - 8 895 123
NM_010628.3 NP_034758.2 7 - 8 900 123
GenBank mRNA AJ132889.1 - 7 - 8 833 123
AK029506.1 - 7 - 8 793 123
AK029776.1 - 7 - 8 865 123
AK087008.1 - 7 - 8 865 123
BC115765.1 - 8 - 9 1093 123

View Full Product Details