Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Afg3l2
Entrez Gene ID
69597
Gene Name
AFG3-like AAA ATPase 2
Gene Aliases
2310036I02Rik, AW260507, Emv66, par
Chromosome Location
Chr.18: 67404760 - 67449136 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027130.1 NP_081406.1 16 - 17 2312 94
GenBank mRNA AK149970.1 - 16 - 17 2284 94
AK151207.1 - 16 - 17 2333 94
AK152115.1 - 16 - 17 2343 94
AK152727.1 - 16 - 17 2343 94
AK164537.1 - 16 - 17 2354 95
AK168668.1 - 16 - 17 2371 94
BC021380.1 - 16 - 17 2309 94
BC022577.1 - 2 - 3 336 94
BC036999.1 - 16 - 17 2356 94
BC043056.1 - 16 - 17 2312 94

View Full Product Details