The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133788.2 | NP_598549.1 | 1 - 2 | 362 | 76 |
NR_037908.1 | 1 - 2 | 362 | 76 | ||
GenBank mRNA | AF209926.1 | - | 1 - 2 | 144 | 76 |
AK154135.1 | - | 1 - 2 | 294 | 76 | |
AK154410.1 | - | 1 - 2 | 276 | 76 | |
BC057132.1 | - | 1 - 2 | 272 | 76 | |
BC110695.1 | - | 1 - 2 | 349 | 76 | |
BY094017.1 | - | 1 - 2 | 362 | 76 |