Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Agbl4
- Entrez Gene ID
-
78933
- Gene Name
- ATP/GTP binding protein-like 4
- Gene Aliases
-
4930578N11Rik, 4931433A01Rik
- Chromosome Location
-
Chr.4: 110397643 - 111664324 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK051109.1 |
- |
3 - 4 |
396 |
79 |
View Full Product Details