Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Add3
Entrez Gene ID
27360
Gene Name
adducin 3 (gamma)
Gene Aliases
AI463285, R75380
Chromosome Location
Chr.19: 53140443 - 53247326 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164099.2 NP_001157571.1 1 - 2 268 72
NM_001164101.2 NP_001157573.1 1 - 2 268 72
GenBank mRNA AK019491.1 - 1 - 2 209 72
AK028481.1 - 1 - 2 173 72
AK035151.1 - 1 - 2 97 72
AK083939.1 - 1 - 2 171 72
BC029196.1 - 1 - 2 133 72

View Full Product Details