The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015099.2 | NP_001015099.2 | 7 - 8 | 859 | 147 |
NM_001167963.1 | NP_001161435.1 | 7 - 8 | 859 | 147 | |
NM_001167964.1 | NP_001161436.1 | 7 - 8 | 859 | 147 | |
GenBank mRNA | AK031333.1 | - | 7 - 8 | 719 | 147 |
AK033513.1 | - | 7 - 8 | 801 | 147 | |
AK084256.1 | - | 7 - 8 | 730 | 147 | |
AK129332.1 | - | 7 - 8 | 762 | 147 | |
BC086455.1 | - | 7 - 8 | 732 | 147 |