Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gltscr1
Entrez Gene ID
243842
Gene Name
glioma tumor suppressor candidate region gene 1
Gene Aliases
-
Chromosome Location
Chr.7: 15970671 - 16048082 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081418.1 NP_001074887.1 9 - 10 3320 81

View Full Product Details