Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oip5
Entrez Gene ID
70645
Gene Name
Opa interacting protein 5
Gene Aliases
5730547N13Rik, Lint-25, Ms18b
Chromosome Location
Chr.2: 119609532 - 119618505 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042653.1 NP_001036118.1 2 - 3 419 94
GenBank mRNA AK045323.1 - 1 - 2 622 94
BC115883.1 - 2 - 3 375 94
BC116660.1 - 2 - 3 376 94

View Full Product Details