Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oip5
Entrez Gene ID
70645
Gene Name
Opa interacting protein 5
Gene Aliases
5730547N13Rik, Lint-25, Ms18b
Chromosome Location
Chr.2: 119609532 - 119618505 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042653.1 NP_001036118.1 3 - 4 539 121
GenBank mRNA AK017825.1 - 3 - 4 504 121
AK045323.1 - 2 - 3 742 121
BC115883.1 - 3 - 4 495 121
BC116660.1 - 3 - 4 496 121

View Full Product Details