Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tubb2a-ps2
Entrez Gene ID
627110
Gene Name
tubulin, beta 2a, pseudogene 2
Gene Aliases
C79445
Chromosome Location
Chr.12: 11882196 - 11882899 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003964.2 1 - 1 189 137
GenBank mRNA BG066514.2 - 1 - 1 189 137


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA C79445.1 - 186 Chr. - - [-] on Build 137


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream RAD51 associated protein 2 Rad51ap2 EG209550 -365.0kb Anti-sense Chr. - 11455759 - 11517161 on Build GRCm38
Downstream family with sequence similarity 49, member A Fam49a 2410157M17Rik, 9630047E15, D12Ertd553e 379.2kb Anti-sense Chr. - 12262129 - 12379058 on Build GRCm38

View Full Product Details