Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spi1
Entrez Gene ID
20375
Gene Name
spleen focus forming virus (SFFV) proviral integration oncogene
Gene Aliases
Dis-1, Dis1, PU.1, Sfpi-1, Sfpi1, Spi-1, Tcfpu1, Tfpu.1
Chromosome Location
Chr.2: 91082381 - 91115757 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011355.1 NP_035485.1 4 - 5 654 95
GenBank mRNA AK142246.1 - 4 - 5 671 95
AK153524.1 - 4 - 5 685 95
AK154945.1 - 4 - 5 706 95
AK155085.1 - 4 - 5 690 95
BC003815.1 - 4 - 5 572 95
L03215.1 - 4 - 5 551 95
M32370.1 - 4 - 5 634 95
M38252.1 - 4 - 5 654 95
X17463.1 - 4 - 5 1342 95

View Full Product Details