Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dennd5a
Entrez Gene ID
19347
Gene Name
DENN/MADD domain containing 5A
Gene Aliases
1500012B19Rik, AJ245569, AW496491, ORF37, Rab6ip1, mKIAA1091
Chromosome Location
Chr.7: 109893780 - 109960458 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021494.1 NP_067469.1 21 - 22 3758 87
GenBank mRNA AJ245569.1 - 20 - 21 3664 87
AK048609.1 - 12 - 13 1412 87
BC022119.1 - 17 - 18 2411 87
BC060230.1 - 21 - 22 3758 87

View Full Product Details