Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekha1
Entrez Gene ID
101476
Gene Name
pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 1
Gene Aliases
AA960558, C920009D07Rik, TAPP1
Chromosome Location
Chr.7: 130865729 - 130913308 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133942.2 NP_598703.1 5 - 6 558 122
GenBank mRNA AK044224.1 - 7 - 8 710 122
AK083340.1 - 7 - 8 732 122
BC020017.1 - 5 - 6 558 122

View Full Product Details