The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081329.1 | NP_001074798.1 | 1 - 2 | 885 | 92 |
GenBank mRNA | AK034954.1 | - | 1 - 2 | 887 | 92 |
AK080486.1 | - | 1 - 2 | 1063 | 92 |