The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081117.2 | NP_001074586.2 | 12 - 13 | 9312 | 118 |
GenBank mRNA | AK133964.1 | - | 1 - 2 | 1836 | 118 |
BC053453.1 | - | 1 - 2 | 1938 | 118 | |
X82786.1 | - | 13 - 14 | 8700 | 118 |