The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009989.3 | NP_034119.1 | 3 - 4 | 370 | 98 |
GenBank mRNA | AK005582.1 | - | 3 - 4 | 304 | 98 |
AK018833.1 | - | 3 - 4 | 304 | 98 | |
AK018851.1 | - | 3 - 4 | 304 | 98 | |
BC049608.1 | - | 3 - 4 | 324 | 98 | |
BC059728.1 | - | 3 - 4 | 366 | 98 | |
M20625.1 | - | 3 - 4 | 365 | 98 | |
X55771.1 | - | 3 - 4 | 303 | 98 |