Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cmya5
Entrez Gene ID
76469
Gene Name
cardiomyopathy associated 5
Gene Aliases
2310076E16Rik, 2310076E21Rik, AA420382, AI504003, Srfsd, Tims, sr553
Chromosome Location
Chr.13: 93040713 - 93145863 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023821.3 NP_076310.2 11 - 12 10831 102
GenBank mRNA AJ245954.1 - 2 - 3 204 102
AJ250188.1 - 2 - 3 204 102
AJ511265.1 - 10 - 11 10508 102
AJ575748.1 - 11 - 12 11020 102
AK010194.1 - 3 - 4 317 102

View Full Product Details