Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Marveld2
Entrez Gene ID
218518
Gene Name
MARVEL (membrane-associating) domain containing 2
Gene Aliases
BC003296, MARVD2, Mrvldc2, Tric, Trica, Tricb, Tricc
Chromosome Location
Chr.13: 100595957 - 100616971 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038602.4 NP_001033691.1 1 - 2 50 71

View Full Product Details