The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163516.2 | NP_001156988.1 | 11 - 12 | 1659 | 81 |
NM_001163517.2 | NP_001156989.1 | 6 - 7 | 781 | 81 | |
NM_001289505.1 | NP_001276434.1 | 13 - 14 | 1640 | 81 | |
NM_021483.4 | NP_067458.2 | 13 - 14 | 1769 | 81 | |
GenBank mRNA | AB032591.1 | - | 13 - 14 | 1764 | 81 |
AK006035.1 | - | 6 - 7 | 782 | 81 | |
AK039764.1 | - | 13 - 14 | 1639 | 81 | |
AK083141.1 | - | 11 - 12 | 1551 | 81 | |
HQ646186.1 | - | 14 - 15 | 1988 | 81 | |
HQ646187.1 | - | 13 - 14 | 1922 | 81 | |
HQ646188.1 | - | 12 - 13 | 1817 | 81 | |
HQ646189.1 | - | 12 - 13 | 1850 | 81 | |
HQ646190.1 | - | 11 - 12 | 1745 | 81 | |
HQ646191.1 | - | 13 - 14 | 2578 | 81 | |
HQ646192.1 | - | 14 - 15 | 1992 | 81 | |
HQ646193.1 | - | 14 - 15 | 1898 | 81 | |
HQ646194.1 | - | 13 - 14 | 1793 | 81 | |
HQ646195.1 | - | 14 - 15 | 1766 | 81 | |
HQ646196.1 | - | 13 - 14 | 1661 | 81 | |
HQ646197.1 | - | 6 - 7 | 781 | 81 |