Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rc3h1
Entrez Gene ID
381305
Gene Name
RING CCCH (C3H) domains 1
Gene Aliases
5730557L09Rik, Gm551, N28103, mKIAA2025
Chromosome Location
Chr.1: 160906411 - 160974976 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024952.2 NP_001020123.1 16 - 17 3261 59
GenBank mRNA AK139648.1 - 16 - 17 3233 59
AK173335.1 - 16 - 17 3207 59
AY948287.1 - 16 - 17 3202 59
BC138663.1 - 15 - 16 2985 59

View Full Product Details