Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zc3h11a
Entrez Gene ID
70579
Gene Name
zinc finger CCCH type containing 11A
Gene Aliases
1110003F06Rik, 5730454B08Rik, G630041M05Rik, Zc3hdc11a, Zc3hh11a, mKIAA0663
Chromosome Location
Chr.1: 133619868 - 133661399 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276767.1 NP_001263696.1 11 - 12 1288 89
NM_144530.6 NP_653113.4 10 - 11 1234 89
GenBank mRNA AK041148.1 - 10 - 11 1208 89
AK078105.1 - 10 - 11 1197 89
AK122341.2 - 9 - 10 4769 89
AK134057.1 - 7 - 8 714 89
AK145740.1 - 10 - 11 1207 89
AK167700.1 - 10 - 11 1208 89
BC005786.1 - 9 - 10 1051 89
BC058552.1 - 9 - 10 1290 89
BC066163.1 - 11 - 12 1246 89
BC066848.1 - 9 - 10 2045 89
FR682665.1 - 11 - 12 1207 89

Gene Symbol
Zbed6
Entrez Gene ID
667118
Gene Name
zinc finger, BED type containing 6
Gene Aliases
Gm8466, MGR
Chromosome Location
Chr.1: 133655879 - 133660885 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FR682664.1 - - 7046 89

View Full Product Details