Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Asb1
- Entrez Gene ID
-
65247
- Gene Name
- ankyrin repeat and SOCS box-containing 1
- Gene Aliases
-
1700029O08Rik, 1700054C17Rik, AI426616, AI851378, mKIAA1146
- Chromosome Location
-
Chr.1: 91539459 - 91559590 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK122456.1 |
- |
1 - 2 |
154 |
106 |
View Full Product Details