The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028339.1 | NP_082615.1 | 1 - 2 | 172 | 68 |
GenBank mRNA | AK013150.1 | - | 1 - 2 | 172 | 68 |
AK030429.1 | - | 1 - 2 | 190 | 68 | |
AK050567.1 | - | 1 - 2 | 171 | 68 | |
AK137059.1 | - | 1 - 2 | 169 | 68 | |
AK150511.1 | - | 1 - 2 | 179 | 68 | |
AK153027.1 | - | 1 - 2 | 175 | 68 | |
AK170468.1 | - | 1 - 2 | 190 | 68 | |
BC017603.1 | - | 1 - 2 | 172 | 68 | |
BC021533.1 | - | 1 - 2 | 193 | 68 |