The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021710.3 | NP_068356.1 | 3 - 4 | 648 | 101 |
GenBank mRNA | AF092093.1 | - | 3 - 4 | 577 | 101 |
AK005283.1 | - | 3 - 4 | 398 | 101 | |
AK178071.1 | - | 2 - 3 | 196 | 101 | |
BC001985.1 | - | 3 - 4 | 648 | 101 | |
BC053339.1 | - | 3 - 4 | 408 | 101 | |
BP426557.1 | - | 1 - 2 | 103 | 101 |