Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyrm7
Entrez Gene ID
75530
Gene Name
LYR motif containing 7
Gene Aliases
1700024C24Rik, 9330147L21Rik
Chromosome Location
Chr.11: 54826866 - 54860907 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029327.3 NP_083603.2 3 - 4 180 155
GenBank mRNA AK006294.1 - 3 - 4 506 155
BC139403.1 - 3 - 4 311 155
BC139406.1 - 3 - 4 311 155
BY776521.1 - 3 - 4 180 155

View Full Product Details