The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029988.2 | NP_001025159.1 | 22 - 23 | 12530 | 89 |
GenBank mRNA | AK132523.1 | - | 5 - 6 | 1753 | 89 |
AK173031.1 | - | 14 - 15 | 4428 | 89 |