The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138660.2 | NP_619601.2 | 8 - 9 | 1582 | 68 |
GenBank mRNA | AF526276.1 | - | 8 - 9 | 1556 | 68 |
AJ292072.1 | - | - | 1796 | 68 | |
AK139608.1 | - | 8 - 9 | 1621 | 68 | |
AK145886.1 | - | 8 - 9 | 1356 | 68 | |
BC034533.1 | - | 5 - 6 | 1226 | 68 | |
BC060672.1 | - | 8 - 9 | 1410 | 68 | |
BC141296.1 | - | 8 - 9 | 1695 | 68 |