Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc59
Entrez Gene ID
98238
Gene Name
leucine rich repeat containing 59
Gene Aliases
AA959742, C78668
Chromosome Location
Chr.11: 94629824 - 94645216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133807.1 NP_598568.1 3 - 4 500 57
GenBank mRNA AK053450.1 - 3 - 4 527 57
AK159374.1 - 3 - 4 556 57
AK159552.1 - 3 - 4 550 57
AK166576.1 - 3 - 4 549 57
AK167604.1 - 3 - 4 491 57
AK169250.1 - 3 - 4 475 57
AK169390.1 - 3 - 4 499 57
AK186256.1 - 1 - 2 73 57
AK196953.1 - - 73 57
BC006877.1 - 3 - 4 500 57
BC066172.1 - 3 - 4 494 57

View Full Product Details