The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130450.1 | NP_001123922.1 | 2 - 3 | 1111 | 66 |
NM_008686.3 | NP_032712.2 | 2 - 3 | 1169 | 66 | |
GenBank mRNA | AK131183.1 | - | 2 - 3 | 1104 | 66 |
AK152040.1 | - | 2 - 3 | 1150 | 66 | |
AK153529.1 | - | 2 - 3 | 1151 | 66 | |
AK154167.1 | - | 2 - 3 | 1080 | 66 | |
AK184212.1 | - | 1 - 2 | 179 | 66 | |
BC043063.1 | - | 1 - 2 | 751 | 66 | |
BC047283.1 | - | 1 - 2 | 511 | 66 | |
BC099895.1 | - | 2 - 3 | 1135 | 66 | |
X78709.1 | - | 1 - 2 | 547 | 66 |